4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
Substrate for alkaline and acid phosphatase determinations
Have Questions?
Catalog NumberQuantity
ALFJ61401.KN100 Pc.
Catalog number ALFJ61401.KN
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 Pc.
Request bulk or custom format
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
Infrared spectrumConforms
Appearance (Form)Pellets
Loss on drying=<1 %
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.
Solubility
It is soluble in water.
Notes
The tablets should be stored -20°C
RUO – Research Use Only
General References:
  1. GN Bowers, RB McComb, A Upretti. 4-nitrophenyl phosphate--characterization of high-purity materials for measuring alkaline phosphatase activity in human serum. Clinical chemistry. 1981, 27, (1), 135-143.
  2. R Rej.; JP Bretaudiere. Effects of metal ions on the measurement of alkaline phosphatase activity. Clinical chemistry. 1980, 26, (3), 423-428.