Acrylamide/Bisacrylamide 37.5:1, powder
Product Image
Thermo Scientific Chemicals
Acrylamide/Bisacrylamide 37.5:1, powder
Premixed powder for preparing nucleic acid PAGE gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
200 g
40 g
Catalog number ALFJ61220.28
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
200 g
Request bulk or custom format
Chemical Identifiers
CAS60558-96-5
IUPAC Name[3-(diethylazaniumyl)propyl]diethylazanium
Molecular FormulaC11H28N2
InChI KeyRCZLVPFECJNLMZ-UHFFFAOYSA-P
SMILESCC[NH+](CC)CCC[NH+](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.4355-1.4405 @ 20°C
Assay (GC)≥96.0%
FormLiquid
Premixed powder for preparing nucleic acid PAGE gels