Acrylamide/Bisacrylamide 37.5:1, powder
Product Image
Thermo Scientific Chemicals
Acrylamide/Bisacrylamide 37.5:1, powder
Premixed powder for preparing nucleic acid PAGE gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
40 g
200 g
Catalog number ALFJ61220.17
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
40 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAcrylamide/Bisacrylamide 37.5:1
Physical FormPowder
Health Hazard 1H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
View more
Premixed powder for preparing nucleic acid PAGE gels