Fluoxetine hydrochloride, 99%, Thermo Scientific Chemicals
Fluoxetine hydrochloride, 99%, Thermo Scientific Chemicals
Fluoxetine hydrochloride, 99%, Thermo Scientific Chemicals
Fluoxetine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fluoxetine hydrochloride, 99%, Thermo Scientific Chemicals
A selective serotonin reuptake inhibitor
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
50 mg
Catalog number ALFJ61197.MD
Price (MYR)
3,107.00
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
3,107.00
Add to cart
Specifications
Chemical Name or MaterialFluoxetine hydrochloride
CAS56296-78-7
Health Hazard 1H302-H315-H318
Health Hazard 2GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Health Hazard 3P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c
View more
Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).
Solubility
Soluble in dimethylsulfoxide and water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Huang, C.; Seip, K. F.; Gjelstad, A.; Shen, X.; Pedersen-Bjergaard, S. Combination of Electromembrane Extraction and Liquid-Phase Microextraction in a Single Step: Simultaneous Group Separation of Acidic and Basic Drugs. Anal. Chem. 2015, 87 (13), 6951-6957.
  2. Chakravarty, P.; Kothari, S.; Deese, A.; Lubach. J. W. Solid-State Characterization of Novel Propylene Glycol Ester Solvates Isolated from Lipid Formulations. Mol. Pharmaceutics. 2015, 12 (7), 2551-2557.