Phosphate-buffered saline (PBS, 1X), sterile-filtered
Phosphate-buffered saline (PBS, 1X), sterile-filtered
Thermo Scientific Chemicals
Phosphate-buffered saline (PBS, 1X), sterile-filtered
Have Questions?
Catalog NumberQuantity
ALFJ61196.AP500 mL
Catalog number ALFJ61196.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Capacity500 mL
Recommended StorageAmbient temperatures
Dilution1 X
Product TypePhosphate Buffered Saline (PBS)
Quantity500 mL
View more
Phosphate-buffered saline (PBS, 1X), sterile is a specifically formulated buffer solution and is used for reconstituting sterile cytokines or antibodies.