Glucose-6-phosphate dehydrogenase
Glucose-6-phosphate dehydrogenase
Thermo Scientific Chemicals
Glucose-6-phosphate dehydrogenase
CAS: 9001-40-5
Have Questions?
Catalog NumberQuantityPackaging
ALFJ61181.4I1 kUPlastic bottle
Catalog number ALFJ61181.4I
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kU
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialGlucose-6-phosphate dehydrogenase
CAS9001-40-5
Recommended StorageStore at -20°C
EINECS Number232-602-6
TSCAYes
View more
Glucose-6-phosphate dehydrogenase, yeast converts p-glucose-6-phosphate to p-glucono-d-lactone-6-phosphate in presence of NADP+ or NAD+ for the determination of blood glucose or creatine kinase. It is used to investigate the effect of dehydroepiandrosterone on cell growth and mitochondrial function in TM-3 cells, in bovine follicular cells and in the incidence of apoptosis and transcript abundance. It is also useful to test ketose reductase activity in developing maize endosperm.