Benzydamine hydrochloride, Thermo Scientific Chemicals
Benzydamine hydrochloride, Thermo Scientific Chemicals
Benzydamine hydrochloride, Thermo Scientific Chemicals
Benzydamine hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzydamine hydrochloride, Thermo Scientific Chemicals
CAS: 132-69-4 | C19H24ClN3O | 345.871 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFJ61180.06
Price (MYR)
374.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
374.00
Add to cart
Specifications
Chemical Name or MaterialBenzydamine hydrochloride
CAS132-69-4
Health Hazard 1H302+H312+H332-H319
Health Hazard 2GHS H Statement
H302-H312-H332-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c
View more
Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Nonsteroidal analgesic and anti-inflammatory compound
  • A prostaglandin synthase inhibitor
RUO – Research Use Only