Sodium phosphate, 0.5M buffer soln., pH 8.5
Sodium phosphate, 0.5M buffer soln., pH 8.5
Thermo Scientific Chemicals
Sodium phosphate, 0.5M buffer soln., pH 8.5
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number ALFJ61151.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS129-73-7
IUPAC Name4-{[4-(dimethylamino)phenyl](phenyl)methyl}-N,N-dimethylaniline
Molecular FormulaC23H26N2
InChI KeyWZKXBGJNNCGHIC-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale green or pale blue
FormCrystals or powder or crystalline powder
CommentMaterial sourced in US and in other countries
Elemental Analysis (typical)Carbon, C: 77.74-89.45% [US from vendor's CofA]
Elemental Analysis (typical)Nitrogen, N: 7.88-9.07% [US from vendor's CofA]