Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals
Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals
Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals
Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerol, Molecular Biology Grade, Thermo Scientific Chemicals
Useful as a cryoprotectant. Glycerol is evolved in the storage and concentration of enzymes. It plays a vital role in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is also used to aid in casting gradient gels, protein stabilizer and storage buffer component.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ALFJ61059.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS9005-12-3
IUPAC Namemethyl(phenyl)silanone
Molecular FormulaC7H8OSi
InChI KeyIXKNJYSAWNJQQY-UHFFFAOYSA-N
SMILESC[Si](=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormViscous liquid
Viscosity450-550 cSt
Infrared spectrumConforms to structure
Useful as a cryoprotectant.Glycerol is evolved in the storage and concentration of enzymes. It plays a vital role in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is also used to aid in casting gradient gels, protein stabilizer and storage buffer component. Further, it is used as a solvent, emollient, pharmaceutical agent and sweetening agent.