Denatonium benzoate, Thermo Scientific Chemicals
Denatonium benzoate, Thermo Scientific Chemicals
Denatonium benzoate, Thermo Scientific Chemicals
Denatonium benzoate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Denatonium benzoate, Thermo Scientific Chemicals
The bitterest compound known
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFJ61048.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6192-52-5
IUPAC Name4-methylbenzene-1-sulfonic acid hydrate
Molecular FormulaC7H10O4S
InChI KeyKJIFKLIQANRMOU-UHFFFAOYSA-N
SMILESO.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=98.5 %
Clarity of solution(c=20, water) Passes test
Heavy metals (as Pb)=<0.001 %
Appearance (Form)Crystalline powder or crystals
View more
The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.
Solubility
Soluble in isopropyl alcohol, methanol. Slightly soluble in acetone. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents. Moisture sensitive.
RUO – Research Use Only
General References:
  1. Kwiatkowski, A.; Czerwicka, M.; Smulko, J.; Stepnowski, P. Detection of Denatonium Benzoate (Bitrex) Remnants in Noncommercial Alcoholic Beverages by Raman Spectroscopy. J. Forensic Sci. 2014, 59 (5), 1358-1363.
  2. Firouzi, M. S.; Firouzi, M.; Nabian, M. H.; Zanjani, L. O.; Zadegan, S. A.; Kamrani, R. S.; Rahimi-Movaghar, V. The effects of picric acid (2,4,6-trinitrophenol) and a bite-deterrent chemical (denatonium benzoate) on autotomy in rats after peripheral nerve lesion. Lab anim 2015, 44 (4), 141-145.