TRIS, 1.0M buffer soln., pH 8.5, 0.2 micron filtered
TRIS, 1.0M buffer soln., pH 8.5, 0.2 micron filtered
TRIS, 1.0M buffer soln., pH 8.5, 0.2 micron filtered
TRIS, 1.0M buffer soln., pH 8.5, 0.2 micron filtered
Thermo Scientific Chemicals
TRIS, 1.0M buffer soln., pH 8.5, 0.2 micron filtered
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61038.AP500 mL
Catalog number ALFJ61038.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTRIS
Name Note0.2 micron filtered
CAS77-86-1
Recommended StorageAmbient temperatures
Concentration1.0 M
View more
TRIS, 1.0M buffer soln., pH 8.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TRIS, 1.0M buffer soln., pH 8.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
Store in cool place. Keep container
RUO – Research Use Only
General References:
  1. DE Soltis.; CH Haufler.; DC Darrow.; GJ Gastony. Starch gel electrophoresis of ferns: a compilation of grinding buffers, gel and electrodebuffers, and staining schedules. American fern journal. 1983, 73 ,(1),9-27.
  2. DE Soltis.; CH Haufler.; DC Darrow.; GJ Gastony. Starch gel electrophoresis of ferns: a compilation of grinding buffers, gel and electrodebuffers, and staining schedules. J. Appl. Cryst.1991, 24 ,409-411.