1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals
1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals
1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals
1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals
Coupling reagent for preparation of polynucleotides
Have Questions?
Catalog NumberQuantity
ALFJ61034.AH200 mL
Catalog number ALFJ61034.AH
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
200 mL
Request bulk or custom format
Chemical Identifiers
CAS7664-39-3
IUPAC Namehydrogen fluoride
Molecular FormulaFH
InChI KeyKRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILESF
View more
SpecificationsSpecification SheetSpecification Sheet
Fluorosilicic acid=<0.01 % (H2SiF6)
Sulfates=<5 ppm (SO4)
Titration with NaOH48 to 51 %
Chloride (Cl)=<5 ppm
Phosphates=<1 ppm (PO4)
View more
1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.
Solubility
Miscible with water.
Notes
Incompatible with reducing agents and alkali metals.
RUO – Research Use Only
General References:
  1. Frija, L. M. T A.; Ismael, A.; Cristiano, M. L. S. Photochemical Transformations of Tetrazole Derivatives: Applications in Organic Synthesis. Molecules 2010, 15 (5), 3757-3774.
  2. Dabbagh, A. H.; Mansoori, Y. New azoic dyes containing (1H)-tetrazole and azido group. Dyes Pigm. 2002, 54 (1), 37-46.