Trypsin Inhibitor, soybeans
Trypsin Inhibitor, soybeans
Thermo Scientific Chemicals
Trypsin Inhibitor, soybeans
Inhibits the pancreatic enzyme trypsin
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
Catalog number ALFJ60982.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Quantity1 g
DescriptionOff-white to Pale Brown
For Use With (Application)Inhibits the pancreatic enzyme trypsin
Recommended StorageKeep cold
CAS9035-81-8
View more
The soybean trypsin inhibitor has been tested for cell culture applications. It has been used in a study to assess the potential application in animal cell culture of an alkaline protease from a non-toxigenic mangrove isolate of Vibri sp. V26. It has also been used in a study to improve the detection of fungi in eosinophilic mucin. It is an inhibitor of trypsin factor Xa, plasmin and neutrophil elastase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dissolve in 1X PBS at 1 to 10mg/ml for stock solution
RUO – Research Use Only
Contents & Storage
2°C to 8°C