Phosphate, 0.5M buffer soln., pH 6.5
Phosphate, 0.5M buffer soln., pH 6.5
Phosphate, 0.5M buffer soln., pH 6.5
Phosphate, 0.5M buffer soln., pH 6.5
Thermo Scientific Chemicals
Phosphate, 0.5M buffer soln., pH 6.5
CAS: 7558-80-7119.976 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60845.AP500 mL
Catalog number ALFJ60845.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS76058-33-8
IUPAC Name2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
Molecular FormulaC17H18N3O2
InChI KeyHBRCDTRQDHMTDA-UHFFFAOYSA-M
SMILESCCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to dark purple powder
FormPowder
Color ChangeRed @ pH 4.5 to yellow @ pH 6.5
lambda-max (UV-Vis)450nm ± 5nm (0.1M NaOH)
Solution Test0.1% in ethanol: clear
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient Temperatures