Sodium phosphate, 0.5M buffer soln., pH 8.0
Sodium phosphate, 0.5M buffer soln., pH 8.0
Thermo Scientific Chemicals
Sodium phosphate, 0.5M buffer soln., pH 8.0
Have Questions?
Catalog NumberQuantity
ALFJ60825.AK250 mL
Catalog number ALFJ60825.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS7726-95-6
IUPAC Namebromide
Molecular FormulaBr
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.80%
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.
Solubility
Soluble in water (121 mg/ml at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.
RUO – Research Use Only
General References:
  1. CR Shaw.; R Prasad. Starch gel electrophoresis of enzymes—a compilation of recipes. Biochemical genetics. 1970, 4,(2), 297-320.
  2. AM Maxam.; W Gilbert. A new method for sequencing DNA. PNAS. 1977, 74,(2), 560-564.