Acrylamide/Bisacrylamide 29:1, powder
Acrylamide/Bisacrylamide 29:1, powder
Thermo Scientific Chemicals
Acrylamide/Bisacrylamide 29:1, powder
Premixed powder for preparing protein DNA gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
40 g
Catalog number ALFJ60824.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS10102-68-8
IUPAC Namecalcium diiodide
Molecular FormulaCaI2
InChI KeyUNMYWSMUMWPJLR-UHFFFAOYSA-L
SMILES[Ca++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤0.01%
Appearance (Color)White
Residual water≤100ppm
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.
Solubility
It is soluble in water.
Notes
Keep away from light. Store at room tempeature.
RUO – Research Use Only
General References:
  1. H Eshaghpour.; DM Crothers. Preparative separation of the complementary strands of DNA restriction fragments by alkaline RPC-5 chromatography. Nucl. Acids Res. 1978, 5,(1), 13-21.
  2. M Bazile, HA Nichols, JA Pojman. Effect of orientation on thermoset frontal polymerization. Journal of Polymer. 2002, 40,(20), 3504-3508.