alpha-Cyclodextrin, 97+%, Thermo Scientific Chemicals
alpha-Cyclodextrin, 97+%, Thermo Scientific Chemicals
alpha-Cyclodextrin, 97+%, Thermo Scientific Chemicals
alpha-Cyclodextrin, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Cyclodextrin, 97+%, Thermo Scientific Chemicals
Useful for selective precipitation of enantiomeric, positional or structural isomers
Have Questions?
Catalog NumberQuantity
ALFJ60687.1425 g
Catalog number ALFJ60687.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS127-63-9
IUPAC Name(benzenesulfonyl)benzene
Molecular FormulaC12H10O2S
InChI KeyKZTYYGOKRVBIMI-UHFFFAOYSA-N
SMILESO=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Identification (FTIR)Conforms
Assay (GC)≥99.0%
FormCrystalline powder or powder
Melting Point (clear melt)122-131?C
Useful for selective precipitation of enantiomeric, positional or structural isomersα-Cyclodextrin is used as a fiber ingredient, an odor or flavor masking agent. It is also useful for emulsification applications. It is also used as whipping fiber and emulsifying fiber. It finds application in medical, healthcare and food and beverage applications. It is also used to lower blood low-density lipoprotein cholesterol levels and lower blood triglyceride levels. It plays an essential role in fat free or fat containing dessert compositions and also employed for the reduction or the replacement of egg white in confectionary and bakery applications. Further, it acts as a supramolecular carrier, complexing agent and controlled drug release. In addition to this, it is used to increase the insulin and leptin sensitivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water at 1%(w/v)
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Guevara, M. A.; Bauer, L. L.; Garleb, K. A.; Fahey, G. C.; de Godoy, M. R. C. Serum lipid profiles, total tract nutrient digestibility, and gastrointestinal tolerance by dogs of alpha-cyclodextrin. J. Anim. Sci. 2015, 93 (5), 2201-2207.
  2. Ma, P.; Liang, F.; Wang, D.; Yang, Q.; Cao, B.; Song, D.; Gao, D.; Wang, X. Selective determination of o-phenylenediamine by surface-enhanced Raman spectroscopy using silver nanoparticles decorated with alpha-cyclodextrin. Microchim. Acta 2015, 182 (1), 167-174.