Phosphate, 0.2M buffer soln., pH 7.0
Phosphate, 0.2M buffer soln., pH 7.0
Phosphate, 0.2M buffer soln., pH 7.0
Phosphate, 0.2M buffer soln., pH 7.0
Thermo Scientific Chemicals
Phosphate, 0.2M buffer soln., pH 7.0
CAS: 7558-80-7119.976 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60669.K21 L
Catalog number ALFJ60669.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Concentration0.2M buffer soln.
Molecular Weight (g/mol)119.976
Quantity1 L
PackagingPlastic Bottle
CAS7558-80-7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient Temperatures