DNAzol™ Reagent
Thermo Scientific Chemicals
DNAzol™ Reagent
DNAzol∣r Reagent, is a complete and ready-to-use electrophoretic reagent to isolate genomic DNA from solid and liquid samples of animal, plant, yeast, and bacterial origin.
Have Questions?
Catalog NumberQuantity
ALFJ60637.AE100 mL
Catalog number ALFJ60637.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS1390-65-4
IUPAC Name3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Molecular FormulaC22H20O13
InChI KeyDGQLVPJVXFOQEV-UHFFFAOYNA-N
SMILESCC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
BSC CertificationPasses
Infrared spectrumConforms
UV spectrumComplies
Appearance (Form)Powder
Appearance (Color)Dark red
DNAzol™ Reagent is used as an electrophoretic reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
  • Cholesterol (cholesterin, cholesteryl alcohol) is a lipid sterol found in vertebrate cell membranes
  • Cholesterol is a cholestanoid, a class of molecule that contains the C-27 cholestane motif
  • A precursor of many steroids and sex hormones in animals including cortisol, aldosterone, testosterone, progesterone, and estrogen
  • It plays a role in neurotransmission and is crucial for organ systems, such as immune, reproductive and endocrine systems
Applications
  • Suitable for in vitro and in vivo applications
  • Stabilized in powder form for laboratory use only
RUO – Research Use Only