Myricetin, 98%, Thermo Scientific Chemicals
Myricetin, 98%, Thermo Scientific Chemicals
Myricetin, 98%, Thermo Scientific Chemicals
Myricetin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Myricetin, 98%, Thermo Scientific Chemicals
CAS: 529-44-2 | C15H10O8 | 318.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
Catalog number ALFJ60450.MF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Chemical Identifiers
CAS1465-25-4
IUPAC Namedihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
Molecular FormulaC12H16Cl2N2
InChI KeyMZNYWPRCVDMOJG-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale grey
Formpowder
Assay (Titration ex Chloride)> 95.0% (dry wt. basis)
Water Content (Karl Fischer Titration)< 5.0%
Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.
Solubility
Soluble in dimethyl sulfoxide, dimethyl formamide and ethanol. Insoluble in water.  
Notes
Store in a cool place. Incompatible with strong oxidizing agents.    
RUO – Research Use Only
General References:
  1. Hong, C.; Xie, Y.; Yao, Y.; Li, G.; Yuan, X.; Shen, H. A novel strategy for pharmaceutical cocrystal generation without knowledge of stoichiometric ratio: myricetin cocrystals and a ternary phase diagram. Pharm. Res. 2015, 32 (1), 47-60.
  2. Iyer, S. C.; Gopal, A.; Halagowder, D. Myricetin induces apoptosis by inhibiting P21 activated kinase 1 (PAK1) signaling cascade in hepatocellular carcinoma. Mol. Cell. Biochem. 2015, 407 (12), 223-237.