Sodium chloride, 5M aq. soln., RNAse free
Sodium chloride, 5M aq. soln., RNAse free
Sodium chloride, 5M aq. soln., RNAse free
Sodium chloride, 5M aq. soln., RNAse free
Thermo Scientific Chemicals
Sodium chloride, 5M aq. soln., RNAse free
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60434.AK250 mL
Catalog number ALFJ60434.AK
Price (MYR)
646.00
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
646.00
Add to cart
Specifications
Chemical Name or MaterialSodium chloride
Name NoteRNAse free
CAS7647-14-5
Recommended StorageAmbient temperatures
Physical FormLiquid
View more
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.
Solubility
Soluble in water (358 mg/ml at 20°C), alcohol (very slightly), glycerol (100 mg/ml), and ammonia. Insoluble in hydrochloric acid.
Notes
Incompatible with strong oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances. Store protected from moisture.
RUO – Research Use Only
General References:
  1. Guangling Song.; Andrej Atrens.; Xianliang Wu.; Bo Zhang. Corrosion behaviour of AZ21, AZ501 and AZ91 in sodium chloride. Corros. Sci.. 1998, 40 (10), 1769-1791.
  2. D. H. Jennings. The Effects Of Sodium Chloride On Higher Plants. Biological Reviews. 1976, 51 (4), 453-486.