Phosphate, 0.2M buffer soln., pH 6.8, Thermo Scientific Chemicals
Phosphate, 0.2M buffer soln., pH 6.8, Thermo Scientific Chemicals
Phosphate, 0.2M buffer soln., pH 6.8, Thermo Scientific Chemicals
Phosphate, 0.2M buffer soln., pH 6.8, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphate, 0.2M buffer soln., pH 6.8, Thermo Scientific Chemicals
Widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
Have Questions?
Catalog NumberQuantity
ALFJ60325.K21 L
Catalog number ALFJ60325.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS97-56-3
IUPAC Name2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline
Molecular FormulaC14H15N3
InChI KeyPFRYFZZSECNQOL-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1N)N=NC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Orange to red-brown to dark brown
Infrared spectrumConforms
Melting point100°C to 104°C
HPLC>=96.0 %
View more
This buffer is widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient Temperatures