N-Methyldopamine hydrochloride, 95%, Thermo Scientific Chemicals
N-Methyldopamine hydrochloride, 95%, Thermo Scientific Chemicals
N-Methyldopamine hydrochloride, 95%, Thermo Scientific Chemicals
N-Methyldopamine hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Methyldopamine hydrochloride, 95%, Thermo Scientific Chemicals
Dopamine agonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFJ60306.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Methyldopamine hydrochloride
CAS62-32-8
ColorWhite
Recommended StorageAmbient temperatures
EINECS Number200-528-3
View more
Andregenic and dopamine receptor agonist; active metabolite of Ibopramine. Also, it is used as internal standard in chromatographic analysis of catecholamine's.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Andregenic and dopamine receptor agonist; active metabolite of Ibopramine. Also, it is used as internal standard in chromatographic analysis of catecholamine′s.
Solubility
Soluble in methanol, water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. A. S. V. Burgen and L. L. Iversen. The inhibition of noradrenaline uptake by sympathomimetic amines in the rat isolated heart. Br J Pharmacol Chemother.1965, 25 (1), 34-49.
  2. P. Lumley and K. J. Broadley. Evidence from agonist and antagonist studies to suggest that the β1-adrenoceptors subserving the positive inotropic and chronotropic responses of the heart do not belong to two separate subgroups. Journal of Pharmacy and Pharmacology. 1977, 29, (1), 598-604.