Tween 20 washing buffer, 0.5% in PBS (10X)
Tween 20 washing buffer, 0.5% in PBS (10X)
Thermo Scientific Chemicals
Tween 20 washing buffer, 0.5% in PBS (10X)
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALFJ60304.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS40046-30-8
IUPAC Name1-isothiocyanato-3,5-dimethylbenzene
Molecular FormulaC9H9NS
InChI KeyDSMXCADWIFIJEX-UHFFFAOYSA-N
SMILESCC1=CC(=CC(C)=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.6185-1.6215 @ 20?C
Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as Western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunologicals, and eventually for incubation solutions of immunoreagents (labeled antibodies) to reduce unspecific background.