Tween 20 washing buffer, 0.5% in PBS (10X)
Tween 20 washing buffer, 0.5% in PBS (10X)
Thermo Scientific Chemicals
Tween 20 washing buffer, 0.5% in PBS (10X)
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ALFJ60304.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS80506-64-5
Molecular FormulaH2NCH2CH2(OCH2CH2)nOCH3
InChI KeyQWCGXANSAOXRFE-UHFFFAOYSA-N
SMILESCOCCOCCN
MDL NumberMFCD00131599
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Proton NMR> 95%
Appearance (Color)Off-white to pale yellow
Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as Western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunologicals, and eventually for incubation solutions of immunoreagents (labeled antibodies) to reduce unspecific background.