Loratadine, 98+%, Thermo Scientific Chemicals
Loratadine, 98+%, Thermo Scientific Chemicals
Loratadine, 98+%, Thermo Scientific Chemicals
Loratadine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Loratadine, 98+%, Thermo Scientific Chemicals
A nonsedating-type histamine H1-receptor antagonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
1 g
Catalog number ALFJ60190.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLoratadine
CAS79794-75-5
Melting Point134°C
ColorWhite
Recommended StorageAmbient temperatures
View more
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is soluble in DMSO (50 mg/ml), ethanol (77 mg/ml at 25°C), water (<1mg/mL at 25°C), chloroform, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Barnett et al. Evaluation of the CNS properties of SCH 29851, a potential non-sedating antihistamine. Agents Actions. 1984, 14,(5-6), 590-7.
  2. Kreutner et al. Antiallergic activity of loratidine, a non-sedating antihistamine. Allergy. 1987, 42,(1), 57-63.