Citrate, 0.5M buffer soln., pH 4.5
Citrate, 0.5M buffer soln., pH 4.5
Citrate, 0.5M buffer soln., pH 4.5
Citrate, 0.5M buffer soln., pH 4.5
Thermo Scientific Chemicals
Citrate, 0.5M buffer soln., pH 4.5
CAS: 6132-04-3294.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number ALFJ60024.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
ColorColorless
Concentration0.5M buffer soln.
Molecular Weight (g/mol)294.10
pH4.5
Quantity500 mL
View more
Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.
Solubility
Fully miscible with water. Insoluble in ethanol.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. H.U. Bergmeyer. Substrate for citrate lyase. ed. Methods of Enzymatic Analysis 3rd ed., 1983, 2173.
  2. D M Ramsay; E P Robertson; E MacArthur. Comparison of anticoagulants for the preservation of prothrombin time specimens. Journal of Clinical Pathology, 1977, 30(8), 766-769.
Contents & Storage
Keep cold