Laemmli SDS sample buffer, reducing (4X)
Laemmli SDS sample buffer, reducing (4X)
Thermo Scientific Chemicals
Laemmli SDS sample buffer, reducing (4X)
Laemmli SDS sample buffer is used to prepare protein samples in the Laemmli SDS-PAGE system.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
25 mL
Catalog number ALFJ60015.AD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS867-44-7
IUPAC Namebis((methylsulfanyl)methanimidamide); sulfuric acid
Molecular FormulaC4H14N4O4S3
InChI KeyBZZXQZOBAUXLHZ-UHFFFAOYSA-N
SMILESCSC(N)=N.CSC(N)=N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White
Laemmli SDS sample buffer is used for protein sample preparation, which is used in the Laemmli SDS-PAGE system. It is a widely used system for protein electrophoresis. Further, it is employed in the polyacrylamide gel analysis of proteins. In addition to this, it is utilized to reduce disulfide bonds in order to denature proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Laemmli SDS sample buffer is composed of several compounds, including 375 mM Tris-HCl (pH 6.8), 9% SDS, 50% glycerol, 6% β-mercaptoethanol, 0.03% bromophenol blue
• The SDS compound present in the Laemmli SDS sample buffer binds noncovalently to proteins. SDS is negatively charged and can mask the intrinsic charge of the protein it binds
Application
• Laemmli SDS sample buffer is used to denature proteins and for protein sample preparation to be used in the SDS-PAGE system
• This sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of the running gel
• It is also used for polyacrylamide protein gel analysis
RUO – Research Use Only
General References:
  1. Lee, G. S.; Kappler, K.; Porter, C. J. H.; Scanlon, M. J.; Nicolazzo, J. A. Fatty Acid Binding Proteins Expressed at the Human Blood-Brain Barrier Bind Drugs in an Isoform-Specific Manner. Pharm. Res. 2015, 32 (10), 3432-3446.
  2. Perot, E.; Enjolras, N.; Le Quellec, S.; Indalecio, A.; Girard, J.; Negrier, C.; Dargaud, Y. Expression and characterization of a novel human recombinant factor IX molecule with enhanced in vitro and in vivo clotting activity. Thromb. Res. 2015, 135 (5), 1017-1024.