Thiamine nitrate, Thermo Scientific Chemicals
Thiamine nitrate, Thermo Scientific Chemicals
Thiamine nitrate, Thermo Scientific Chemicals
Thiamine nitrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiamine nitrate, Thermo Scientific Chemicals
CAS: 532-43-4 | C12H17N5O4S | 327.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Catalog number ALFJ60014.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6915-15-7
IUPAC Name2-hydroxybutanedioic acid
Molecular FormulaC4H6O5
InChI KeyBJEPYKJPYRNKOW-UHFFFAOYNA-N
SMILESOC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
Optical Rotation-0.5? to +0.5? (c= 8.5 in water)
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%