Thiamine nitrate, Thermo Scientific Chemicals
Thiamine nitrate, Thermo Scientific Chemicals
Thiamine nitrate, Thermo Scientific Chemicals
Thiamine nitrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiamine nitrate, Thermo Scientific Chemicals
CAS: 532-43-4 | C12H17N5O4S | 327.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Catalog number ALFJ60014.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS144222-34-4
IUPAC NameN-[(1R,2R)-2-azaniumyl-1,2-diphenylethyl]-4-methylbenzene-1-sulfonamide
Molecular FormulaC21H23N2O2S
InChI KeyUOPFIWYXBIHPIP-NHCUHLMSSA-O
SMILESCC1=CC=C(C=C1)S(=O)(=O)N[C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)125.0-131.0°C
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream