Thiabendazole, 98+%, Thermo Scientific Chemicals
Thiabendazole, 98+%, Thermo Scientific Chemicals
Thiabendazole, 98+%, Thermo Scientific Chemicals
Thiabendazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiabendazole, 98+%, Thermo Scientific Chemicals
Fungicide and antihelminthic
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Catalog number ALFJ60009.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThiabendazole
CAS148-79-8
Melting Point304°C to 305°C
ColorWhite
Recommended StorageAmbient temperatures
View more
Thiabendazole, a metal chelator and helminth-specific fumarate reductase inhibitor, is used primarily as an anthelmintic to control infections by helminth organisms. It is also used to control mold and fungal growth in plants. Also used to treat parasitic infections in humans. It is used clinically to treat threadworm, cutaneous larva migrans, visceral larva migrans, and trichinosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in methanol and dimethyl sulfoxide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Roslyn S Thelingwani; Simbarashe P Zvada; Hugues Dolgos; Anna-Lena B Ungell; Collen M Masimirembwa. In vitro and in silico identification and characterization of thiabendazole as a mechanism-based inhibitor of CYP1A2 and simulation of possible pharmacokinetic drug-drug interactions. Drug Metabolism and Disposition, 2009, 37(6), 1286-1294.
  2. Craig Knox et. al. DrugBank 3.0: a comprehensive resource for 'omics' research on drugs. Nucleic Acids Research, 2012, 39(3), D1035-D1041.