N-Isopropylacrylamide, 97%, Thermo Scientific Chemicals
N-Isopropylacrylamide, 97%, Thermo Scientific Chemicals
N-Isopropylacrylamide, 97%, Thermo Scientific Chemicals
N-Isopropylacrylamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Isopropylacrylamide, 97%, Thermo Scientific Chemicals
CAS: 2210-25-5 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ALFH66262.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight113.16
Unit Size
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Firdaus Yhaya; Sandra Binauld; Yoseop Kim; Martina H Stenzel. Shell cross-linking of cyclodextrin-based micelles via supramolecular chemistry for the delivery of drugs. Macromolecular Rapid Communications.201233(21), 1868-1874.
  2. R Yoshida. [Controlled release using temperature-responsive poly(N-isopropylacrylamide) gel]. Japanese journal of clinical medicine.199856(3), 670-674.