2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number ALFH66063.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Formgranules
Appearance (Color)Black
Iodine value≥ 1100
Water Content (Karl Fischer Titration)≤ 5.0%
Particle Size≤ 0.5% is < 0.60mm