2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number ALFH66063.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone
CAS17720-60-4
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c