Methyl 2-bromomethyl-3-nitrobenzoate, 95%, Thermo Scientific™
Methyl 2-bromomethyl-3-nitrobenzoate, 95%, Thermo Scientific™
Methyl 2-bromomethyl-3-nitrobenzoate, 95%, Thermo Scientific™
Methyl 2-bromomethyl-3-nitrobenzoate, 95%, Thermo Scientific™
Methyl 2-bromomethyl-3-nitrobenzoate, 95%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH65351.031 g
Catalog number ALFH65351.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl 2-bromomethyl-3-nitrobenzoate
CAS98475-07-1
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
Recommended StorageAmbient temperatures
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.