2-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
2-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
2-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
2-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
CAS: 28320-31-2 | C15H13Br | 273.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFH64732.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS2233-18-3
IUPAC Name4-hydroxy-3,5-dimethylbenzaldehyde
Molecular FormulaC9H10O2
InChI KeyUYGBSRJODQHNLQ-UHFFFAOYSA-N
SMILESCC1=CC(C=O)=CC(C)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or yellow to orange or pale brown
FormCrystals or Crystalline powder
Assay (GC)≥97.0%
Melting Point (clear melt)111.5-117.5?C
This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Wei Wei; Peter I. Djurovich; and Mark E. Thompson. Properties of Fluorenyl Silanes in Organic Light Emitting Diodes. Chem. Mater.2010 22(5), 1724-1731.
  2. Cai-Jun Zheng; Wei-Ming Zhao; Zhi-Qiang Wang; Da Huang; Jun Ye; Xue-Mei Ou; Xiao-Hong Zhang; Chun-Sing Leec; and Shuit-Tong Lee. Highly efficient non-doped deep-blue organic light-emitting diodes based on anthracene derivatives. J. Mater. Chem.2010 20(8), 1560-1566.