4-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
4-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
4-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
4-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
4-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH64639.065 g
Catalog number ALFH64639.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS539-82-2
IUPAC Nameethyl pentanoate
Molecular FormulaC7H14O2
InChI KeyICMAFTSLXCXHRK-UHFFFAOYSA-N
SMILESCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3990 to 1.4010 (20°C, 589 nm)
GC>=98.5 %