3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals
3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals
3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals
3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH64546.031 g
Catalog number ALFH64546.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material3-(Aminomethyl)benzoic acid hydrochloride
CAS876-03-9
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only
F. Markwardt.; H. Landmann.; P. Walsmann. Comparative Studies on the Inhibition of Trypsin, Plasmin, and Thrombin by Derivatives of Benzylamine and Benzamidine. European Journal of Biochemistry. 1968, 6 (4),502-506 .