Nortropinone hydrochloride, 97%, Thermo Scientific Chemicals
Nortropinone hydrochloride, 97%, Thermo Scientific Chemicals
Nortropinone hydrochloride, 97%, Thermo Scientific Chemicals
Nortropinone hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nortropinone hydrochloride, 97%, Thermo Scientific Chemicals
CAS: 25602-68-0 | C7H12ClNO | 161.629 g/mol
Have Questions?
Catalog NumberQuantity
ALFH64485.065 g
Catalog number ALFH64485.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS14459-95-1
IUPAC Nametetrapotassium hexacyanoironbis(ylium) trihydrate
Molecular FormulaC6H6FeK4N6O3
InChI KeyNLYKFUFIUWPPIZ-UHFFFAOYSA-N
SMILESO.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Titration with KMnO498.5 to 102.0 %
Insoluble matter=<0.005 %
Sulfate (SO4)Passes test (limit about 0.01%)