1-Indoleacetic acid, 95%, Thermo Scientific Chemicals
1-Indoleacetic acid, 95%, Thermo Scientific Chemicals
1-Indoleacetic acid, 95%, Thermo Scientific Chemicals
1-Indoleacetic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Indoleacetic acid, 95%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number ALFH63935.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7784-26-1
IUPAC Namealuminium(3+) ammonium dodecahydrate disulfate
Molecular FormulaAlH28NO20S2
InChI KeyWZUKKIPWIPZMAS-UHFFFAOYSA-K
SMILES[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥99.0%