1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number ALFH62185.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Boc-N-Fmoc-D-tryptophan
CAS163619-04-3
Recommended StorageKeep cold; Store under Nitrogen
TSCANo
Molecular FormulaC31H30N2O6