1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number ALFH62185.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS766-85-8
IUPAC Name1-iodo-3-methoxybenzene
Molecular FormulaC7H7IO
InChI KeyRSHBAGGASAJQCH-UHFFFAOYSA-N
SMILESCOC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Liquid
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=96.0 %