2-Amino-5-bromo-2'-fluorobenzophenone, 95%, Thermo Scientific Chemicals
2-Amino-5-bromo-2'-fluorobenzophenone, 95%, Thermo Scientific Chemicals
2-Amino-5-bromo-2'-fluorobenzophenone, 95%, Thermo Scientific Chemicals
2-Amino-5-bromo-2'-fluorobenzophenone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-bromo-2'-fluorobenzophenone, 95%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH61882.065 g
Catalog number ALFH61882.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS50-01-1
IUPAC Namehydrogen guanidine chloride
Molecular FormulaCH6ClN3
InChI KeyPJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Water=<0.3 %
Anticaking agent=<0.4 % (SiO2)
Appearance (Color)White to light yellow
Melting point179°C to 186°C