3-(Bromomethyl)thiophene, 97%, Thermo Scientific Chemicals
3-(Bromomethyl)thiophene, 97%, Thermo Scientific Chemicals
3-(Bromomethyl)thiophene, 97%, Thermo Scientific Chemicals
3-(Bromomethyl)thiophene, 97%, Thermo Scientific Chemicals
3-(Bromomethyl)thiophene, 97%, Thermo Scientific Chemicals
CAS: 34846-44-1 | C5H5BrS | 177.06 g/mol
Have Questions?
Catalog NumberQuantity
ALFH61497.031 g
Catalog number ALFH61497.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS10031-51-3
IUPAC Nameerbium(3+) pentahydrate trinitrate
Molecular FormulaErH10N3O14
InChI KeyLWHHUEHWVBVASY-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
Appearance (Form)Crystalline powder or crystals
Assay33 to 39 % (Er)
Total conc. of imp.as REO=<0.1 %