Bis(2-[di(1-adamantyl)phosphino]ethyl)amine, 97+%, Thermo Scientific Chemicals
Bis(2-[di(1-adamantyl)phosphino]ethyl)amine, 97+%, Thermo Scientific Chemicals
Bis(2-[di(1-adamantyl)phosphino]ethyl)amine, 97+%, Thermo Scientific Chemicals
Bis(2-[di(1-adamantyl)phosphino]ethyl)amine, 97+%, Thermo Scientific Chemicals
Bis(2-[di(1-adamantyl)phosphino]ethyl)amine, 97+%, Thermo Scientific Chemicals
CAS: 1086138-36-4 | C44H69NP2 | 673.99 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60853.MD250 mg
Catalog number ALFH60853.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS2244-16-8
IUPAC Name(5S)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
Molecular FormulaC10H14O
InChI KeyULDHMXUKGWMISQ-VIFPVBQESA-N
SMILESCC(=C)[C@H]1CC=C(C)C(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4960 to 1.5010 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents. It is sensitive to air.
RUO – Research Use Only