Imidazolidinyl urea, Thermo Scientific Chemicals
Imidazolidinyl urea, Thermo Scientific Chemicals
Imidazolidinyl urea, Thermo Scientific Chemicals
Imidazolidinyl urea, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Imidazolidinyl urea, Thermo Scientific Chemicals
CAS: 39236-46-9 | C11H16N8O8 | 388.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFH60729.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS79-43-6
IUPAC Name2,2-dichloroacetic acid
Molecular FormulaC2H2Cl2O2
InChI KeyJXTHNDFMNIQAHM-UHFFFAOYSA-N
SMILESOC(=O)C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<100 APHA
Infrared spectrumConforms
Refractive index1.4650 to 1.4670 (20°C, 589 nm)
Titration with NaOH99.0 to 101.0 %