Imidazolidinyl urea, Thermo Scientific Chemicals
Imidazolidinyl urea, Thermo Scientific Chemicals
Imidazolidinyl urea, Thermo Scientific Chemicals
Imidazolidinyl urea, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Imidazolidinyl urea, Thermo Scientific Chemicals
CAS: 39236-46-9 | C11H16N8O8 | 388.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFH60729.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialImidazolidinyl urea
CAS39236-46-9
Health Hazard 1H317
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3P261-P272-P280g-P302+P352-P333+P313-P363-P501c