Bis(trimethylsiloxy)methylsilane, 97%, Thermo Scientific Chemicals
Bis(trimethylsiloxy)methylsilane, 97%, Thermo Scientific Chemicals
Bis(trimethylsiloxy)methylsilane, 97%, Thermo Scientific Chemicals
Bis(trimethylsiloxy)methylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(trimethylsiloxy)methylsilane, 97%, Thermo Scientific Chemicals
CAS: 1873-88-7 | C7H21O2Si3 | 221.498 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60586.AD50 mL
Catalog number ALFH60586.AD
Price (MYR)
755.00
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (MYR)
755.00
Add to cart
Specifications
Boiling Point142°C
Chemical Name or MaterialBis(trimethylsiloxy)methylsilane
CAS1873-88-7
Health Hazard 1H225
Health Hazard 2GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.
Solubility
Miscible with acetone, ethanol, diethyl ether. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
RUO – Research Use Only
Qingguo Wu; Karen K. Gleason. Plasma-Enhanced CVD of Organosilicate Glass (OSG) Films Deposited from Octamethyltrisiloxane, Bis(trimethylsiloxy)methylsilane, and 1,1,3,3-Tetramethyldisiloxane. Plasmas and Polymers.2003, 8 31-41.