Oleanolic acid, 97%
Oleanolic acid, 97%
Oleanolic acid, 97%
Thermo Scientific Chemicals
Oleanolic acid, 97%
CAS: 508-02-1 | C30H48O3 | 456.711 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60445.MD250 mg
Catalog number ALFH60445.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS589-15-1
IUPAC Name1-bromo-4-(bromomethyl)benzene
Molecular FormulaC7H6Br2
InChI KeyYLRBJYMANQKEAW-UHFFFAOYSA-N
SMILESBrCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥98.0% (GC)
FormPowder
Melting Point62.0-64.0°C