Dicumyl peroxide, 98%, Thermo Scientific Chemicals
Dicumyl peroxide, 98%, Thermo Scientific Chemicals
Dicumyl peroxide, 98%, Thermo Scientific Chemicals
Dicumyl peroxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dicumyl peroxide, 98%, Thermo Scientific Chemicals
CAS: 80-43-3 | C18H22O2 | 270.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFH60442.36
Price (MYR)
1,193.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,193.00
Add to cart
Specifications
Chemical Name or MaterialDicumyl peroxide
CAS80-43-3
Health Hazard 1H242-H315-H319
Health Hazard 2GHS H Statement
H242-H315-H319
Heating may cause a fire.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P210-P220-P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c
View more
Dicumyl peroxide (DCP) is used:
  • in vulcanization of rubber.
  • as a cross linking agent in the synthesis of polylactic acid composite fibers.
  • in the preparation of polyethylene composites.
  • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dicumyl peroxide (DCP) is used: • in vulcanization of rubber. • as a cross linking agent in the synthesis of polylactic acid composite fibers. • in the preparation of polyethylene composites. • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.
Solubility
Soluble in chloroform and most organic solvents. Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents and acids.
RUO – Research Use Only
General References:
  1. Ray A. Dickie.; John D. Ferry. Dynamic Mechanical Properties of Cross-Linked Rubbers. III. Dicumyl Peroxide Vulcanizates of Natural Rubber1. J. Phys. Chem. 1966, 70 (8),2594-2600.
  2. S. Sapieha.; P. Allard.; Y. H. Zang. Dicumyl peroxide-modified cellulose/LLDPE composites. J. Appl. Polym. Sci. 1990, 41 (9-10),2039-2048.